Assay Details
| Gene Symbol: SNX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016224.4 | NP_057308.1 | 1 - 2 | 280 | 118 | |
| GenBank mRNA | AF172847.1 | - | 1 - 2 | 160 | 117 | |
| AK314360.1 | - | 1 - 2 | 114 | 118 | ||
| BC005022.1 | - | 1 - 2 | 187 | 3832234 | 118 | |
| BG766499.1 | - | 1 - 2 | 190 | 118 | ||
| DA315945.1 | - | 1 - 2 | 281 | 118 | ||
| DA336311.1 | - | 1 - 2 | 226 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001126.hg
Additional Information:
This assay is in the 5'UTR region of the AF172847.1,AK314360.1,BC005022.1,NM_016224.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
intracellular protein transport
endocytosis
receptor-mediated endocytosis
mitotic nuclear division
vesicle organization
endosomal transport
positive regulation of protein oligomerization
cleavage furrow formation
positive regulation of GTPase activity
positive regulation of protein kinase activity
positive regulation of membrane protein ectodomain proteolysis
lipid tube assembly
membrane tubulation
cell-cell adhesion
protein binding
1-phosphatidylinositol binding
ubiquitin protein ligase binding
phosphatidylinositol binding
protein homodimerization activity
Arp2/3 complex binding
cadherin binding involved in cell-cell adhesion
mitotic cytokinesis
intracellular protein transport
endocytosis
receptor-mediated endocytosis
mitotic nuclear division
vesicle organization
endosomal transport
positive regulation of protein oligomerization
cleavage furrow formation
positive regulation of GTPase activity
positive regulation of protein kinase activity
positive regulation of membrane protein ectodomain proteolysis
lipid tube assembly
membrane tubulation
cell-cell adhesion
protein binding
1-phosphatidylinositol binding
ubiquitin protein ligase binding
phosphatidylinositol binding
protein homodimerization activity
Arp2/3 complex binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06013381.hg.1 | TC06001126.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013381.hg.1 | ENST00000392185.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013381.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013382.hg.1 | TC06001126.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013382.hg.1 | ENST00000392185.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013382.hg.1 | ENST00000539592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013382.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013384.hg.1 | TC06001126.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013384.hg.1 | ENST00000392185.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013384.hg.1 | ENST00000539592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013384.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013383.hg.1 | TC06001126.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013383.hg.1 | ENST00000392185.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013383.hg.1 | ENST00000539592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013383.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199777.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199777.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199777.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199777.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199777.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199777.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199777.hg.1 | TC0600014196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600199774.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199774.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199774.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199774.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199774.hg.1 | TC0600014196.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000392185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | ENST00000611124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014196.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199773.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199773.hg.1 | NM_016224.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199773.hg.1 | TC0600014196.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 107212 | - | - |

SNX9
Genomic Map