Assay Details
| Gene Symbol: MED12L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053002.5 | NP_443728.3 | 19 - 20 | 2979 | 100 | |
| XM_006713487.3 | XP_006713550.1 | 21 - 22 | 3356 | 100 | ||
| XM_011512390.2 | XP_011510692.1 | 20 - 21 | 3251 | 100 | ||
| XM_011512394.2 | XP_011510696.1 | 21 - 22 | 3356 | 100 | ||
| XM_011512399.2 | XP_011510701.1 | 21 - 22 | 3356 | 100 | ||
| XM_017005676.1 | XP_016861165.1 | 21 - 22 | 3356 | 100 | ||
| XM_017005678.1 | XP_016861167.1 | 21 - 22 | 3356 | 100 | ||
| XM_017005679.1 | XP_016861168.1 | 20 - 21 | 3086 | 100 | ||
| XM_017005680.1 | XP_016861169.1 | 19 - 20 | 3074 | 100 | ||
| XM_017005681.1 | XP_016861170.1 | 6 - 7 | 742 | 100 | ||
| GenBank mRNA | AB046855.1 | - | 6 - 7 | 722 | 100 | |
| AF399708.1 | - | 19 - 20 | 2888 | 100 | ||
| BC144559.1 | - | 17 - 18 | 2463 | 9053087 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000363124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000480026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000480026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000488092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000488092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |

MED12L
Genomic Map