Assay Details
| Gene Symbol: SLC40A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014585.5 | NP_055400.1 | 5 - 6 | 861 | 78 | |
| XM_005246505.1 | XP_005246562.1 | 6 - 7 | 996 | 78 | ||
| XM_017003938.1 | XP_016859427.1 | 6 - 7 | 1110 | 78 | ||
| GenBank mRNA | AF215636.1 | - | 5 - 6 | 814 | 78 | |
| AF226614.1 | - | 5 - 6 | 814 | 78 | ||
| AF231121.1 | - | 5 - 6 | 814 | 78 | ||
| AK223236.1 | - | 5 - 6 | 836 | 78 | ||
| AK292146.1 | - | 5 - 6 | 836 | 78 | ||
| AK314827.1 | - | 5 - 6 | 836 | 78 | ||
| AL136944.1 | - | 5 - 6 | 824 | 78 | ||
| BC035893.1 | - | 5 - 6 | 825 | 5213437 | 78 | |
| BC037733.1 | - | 5 - 6 | 940 | 4823308 | 78 | |
| CR533564.1 | - | 5 - 6 | 510 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002614.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
lymphocyte homeostasis
endothelium development
cellular iron ion homeostasis
regulation of transcription from RNA polymerase II promoter in response to iron
iron ion transmembrane transport
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
spleen trabecula formation
multicellular organismal iron ion homeostasis
ferrous iron export
receptor activity
iron ion transmembrane transporter activity
protein binding
ferrous iron transmembrane transporter activity
peptide hormone binding
iron channel activity
lymphocyte homeostasis
endothelium development
cellular iron ion homeostasis
regulation of transcription from RNA polymerase II promoter in response to iron
iron ion transmembrane transport
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
spleen trabecula formation
multicellular organismal iron ion homeostasis
ferrous iron export
receptor activity
iron ion transmembrane transporter activity
protein binding
ferrous iron transmembrane transporter activity
peptide hormone binding
iron channel activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02041842.hg.1 | TC02002614.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041842.hg.1 | ENST00000261024.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041842.hg.1 | ENST00000481497.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041842.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041843.hg.1 | TC02002614.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041843.hg.1 | ENST00000261024.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041843.hg.1 | ENST00000481497.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02041843.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427419.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427419.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427419.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427419.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000427419.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000440626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000440626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000440626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000440626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211404.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211404.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211404.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211404.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211404.hg.1 | TC0200015211.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000418714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427419.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427419.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427419.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427419.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000427419.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000440626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000440626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000440626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000440626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000455320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | ENST00000479598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015211.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211405.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211405.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211405.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211405.hg.1 | ENST00000261024.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211405.hg.1 | ENST00000427241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200211405.hg.1 | NM_014585.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200211405.hg.1 | TC0200015211.hg.1 | Clariom D TC |

SLC40A1
Genomic Map