Assay Details
| Gene Symbol: ZNHIT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170670.1 | NP_001164141.1 | 3 - 4 | 753 | 72 | |
| NM_017953.3 | NP_060423.3 | 2 - 3 | 870 | 72 | ||
| XM_011541614.2 | XP_011539916.1 | 2 - 3 | 884 | 72 | ||
| GenBank mRNA | AF308296.1 | - | 2 - 3 | 517 | 72 | |
| AK000736.1 | - | 2 - 3 | 242 | 72 | ||
| AK000767.1 | - | 2 - 3 | 779 | 72 | ||
| AK298149.1 | - | 3 - 4 | 753 | 72 | ||
| AK314566.1 | - | 2 - 3 | 798 | 72 | ||
| BC026236.1 | - | 2 - 3 | 857 | 4664103 | 72 | |
| BC110898.1 | - | 2 - 3 | 788 | 6012059 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002831.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044066.hg.1 | TC01002831.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044066.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044066.hg.1 | ENST00000431532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044066.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044066.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210143.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210143.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210143.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210143.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210143.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210143.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210143.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210143.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210143.hg.1 | TC0100014788.hg.1 | Clariom D TC |

ZNHIT6
Genomic Map