Assay Details
| Gene Symbol: ZNHIT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170670.1 | NP_001164141.1 | 9 - 10 | 1266 | 143 | |
| NM_017953.3 | NP_060423.3 | 8 - 9 | 1383 | 143 | ||
| GenBank mRNA | AK000736.1 | - | 8 - 9 | 755 | 143 | |
| AK000767.1 | - | 8 - 9 | 1292 | 143 | ||
| AK298149.1 | - | 9 - 10 | 1266 | 143 | ||
| AK314566.1 | - | 8 - 9 | 1311 | 143 | ||
| AL442074.1 | - | 8 - 9 | 1368 | 143 | ||
| BC026236.1 | - | 8 - 9 | 1370 | 4664103 | 143 | |
| BC110898.1 | - | 8 - 9 | 1301 | 6012059 | 143 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002831.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01044060.hg.1 | TC01002831.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044060.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044060.hg.1 | ENST00000431532.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044060.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044060.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210137.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210137.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210137.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210137.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210137.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210137.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210137.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210137.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210137.hg.1 | TC0100014788.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000370574.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | ENST00000431532.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_001170670.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014788.hg.1 | NM_017953.3 | Refseq |

ZNHIT6
Genomic Map