Assay Details
| Gene Symbol: TMEM87B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032824.2 | NP_116213.1 | 7 - 8 | 1019 | 98 | |
| XM_005263827.2 | XP_005263884.1 | 7 - 8 | 973 | 98 | ||
| XM_006712807.2 | XP_006712870.1 | 7 - 8 | 976 | 98 | ||
| XM_017005121.1 | XP_016860610.1 | 5 - 6 | 380 | 98 | ||
| GenBank mRNA | AK027587.1 | - | 7 - 8 | 1004 | 98 | |
| AK074243.1 | - | 4 - 5 | 277 | 98 | ||
| AK290294.1 | - | 7 - 8 | 662 | 98 | ||
| AY429534.1 | - | 5 - 6 | 380 | 98 | ||
| AY429535.1 | - | 6 - 7 | 471 | 98 | ||
| BC115373.1 | - | 7 - 8 | 820 | 40026807 | 98 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000703.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010847.hg.1 | TC02000703.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010847.hg.1 | ENST00000283206.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010847.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010846.hg.1 | TC02000703.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010846.hg.1 | ENST00000283206.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02010846.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162136.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162136.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162136.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162136.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162136.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162136.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162136.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162136.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162136.hg.1 | TC0200008900.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200162137.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162137.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162137.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162137.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162137.hg.1 | TC0200008900.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000283206.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000452614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000463427.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | ENST00000471632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008900.hg.1 | NM_032824.2 | Refseq |

TMEM87B
Genomic Map