Assay Details
| Gene Symbol: STK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005259618.3 | XP_005259675.1 | 8 - 9 | 1731 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STK11| Gene Symbol: STK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005259618.3 | XP_005259675.1 | 8 - 9 | 1731 | 104 | |
Set Membership: |
|