| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000309794.15 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000374611.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000390006.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000412079.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000457415.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000577410.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000577432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000577812.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000579198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000580296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000580748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000581202.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000581743.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000581795.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000582907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000584411.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
NM_001038618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
NM_012336.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
NM_031968.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000309794.15 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000374611.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000390006.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000412079.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000457415.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000577410.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000577432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000577812.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000579198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000580296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000580748.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000581202.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000581743.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000581795.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000582907.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
ENST00000584411.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
NM_001038618.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
NM_012336.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
NM_031968.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700170357.hg.1 |
TC1700009256.hg.1 |
Clariom D TC |