Assay Details
| Gene Symbol: PEX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000318.2 | NP_000309.1 | 3 - 4 | 440 | 64 | |
| NM_001079867.1 | NP_001073336.1 | 2 - 3 | 408 | 64 | ||
| NM_001172086.1 | NP_001165557.1 | 4 - 5 | 446 | 64 | ||
| GenBank mRNA | BC000661.2 | - | 3 - 4 | 213 | 3347824 | 64 |
| BC005375.1 | - | 2 - 3 | 209 | 3934225 | 64 | |
| BC093043.1 | - | 3 - 4 | 226 | 30399827 | 64 | |
| DA210382.1 | - | 3 - 4 | 440 | 64 | ||
| M85038.1 | - | 3 - 4 | 247 | 64 | ||
| M86852.1 | - | 3 - 4 | 355 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001347.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC000661.2,BC005375.1,BC093043.1,M85038.1,M86852.1,NM_000318.2,NM_001079867.1,NM_001172086.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
very long-chain fatty acid metabolic process
negative regulation of transcription from RNA polymerase II promoter
fatty acid beta-oxidation
peroxisome organization
protein import into peroxisome matrix
protein destabilization
negative regulation of fibroblast proliferation
negative regulation of epithelial cell proliferation
protein binding
zinc ion binding
very long-chain fatty acid metabolic process
negative regulation of transcription from RNA polymerase II promoter
fatty acid beta-oxidation
peroxisome organization
protein import into peroxisome matrix
protein destabilization
negative regulation of fibroblast proliferation
negative regulation of epithelial cell proliferation
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08017331.hg.1 | TC08001347.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017331.hg.1 | ENST00000357039.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017331.hg.1 | ENST00000419564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017331.hg.1 | ENST00000520103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017331.hg.1 | ENST00000522527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017331.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017331.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017331.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017331.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000518986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000518986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000518986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000520203.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017328.hg.1 | TC08001347.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017328.hg.1 | ENST00000357039.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017328.hg.1 | ENST00000419564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017328.hg.1 | ENST00000520103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017328.hg.1 | ENST00000522527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017328.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017328.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017328.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08017328.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | ENST00000518986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | ENST00000518986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800173775.hg.1 | TC0800010869.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000357039.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000518986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000518986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000518986.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000519956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520103.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000520203.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | ENST00000522527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_000318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001079867.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172086.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010869.hg.1 | NM_001172087.1 | Refseq |

PEX2
Genomic Map