Assay Details
| Gene Symbol: CAPN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014296.2 | NP_055111.1 | 8 - 9 | 1211 | 80 | |
| XM_011533555.2 | XP_011531857.1 | 8 - 9 | 1062 | 80 | ||
| XM_011533556.2 | XP_011531858.1 | 9 - 10 | 1294 | 80 | ||
| XM_011533557.2 | XP_011531859.1 | 8 - 9 | 1184 | 80 | ||
| XM_017006043.1 | XP_016861532.1 | 9 - 10 | 1294 | 80 | ||
| XM_017006044.1 | XP_016861533.1 | 8 - 9 | 1184 | 80 | ||
| XM_017006045.1 | XP_016861534.1 | 9 - 10 | 1250 | 80 | ||
| GenBank mRNA | AB028639.1 | - | 8 - 9 | 1192 | 80 | |
| AK314253.1 | - | 8 - 9 | 1173 | 80 | ||
| BC056202.1 | - | 8 - 9 | 1189 | 5274889 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000092.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002016.hg.1 | TC03000092.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002016.hg.1 | ENST00000253693.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03002016.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146776.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146776.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146776.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146776.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146776.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146776.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146776.hg.1 | TC0300006702.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000253693.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000415565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000418994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000443567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000457023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000463417.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | ENST00000472400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006702.hg.1 | NM_014296.2 | Refseq |

CAPN7
Genomic Map