Assay Details
| Gene Symbol: PKN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317926.1 | NP_001304855.1 | 14 - 15 | 2133 | 82 | |
| NM_013355.4 | NP_037487.2 | 14 - 15 | 2133 | 82 | ||
| XM_005251946.3 | XP_005252003.1 | 14 - 15 | 2688 | 82 | ||
| XM_006717080.2 | XP_006717143.1 | 14 - 15 | 2202 | 82 | ||
| XM_017014649.1 | XP_016870138.1 | 14 - 15 | 2204 | 82 | ||
| XM_017014650.1 | XP_016870139.1 | 14 - 15 | 2342 | 82 | ||
| GenBank mRNA | AB019692.1 | - | 14 - 15 | 1721 | 82 | |
| BC062558.1 | - | 14 - 15 | 1736 | 6649806 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000699.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008634.hg.1 | TC09000699.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008634.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008634.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159870.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159870.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159870.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900159870.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900159870.hg.1 | TC0900008881.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000291906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000483521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | ENST00000485301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008881.hg.1 | NM_013355.4 | Refseq |

PKN3
Genomic Map