| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008619.hg.1 |
TC09000699.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008619.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09008619.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000483521.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
ENST00000485301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008881.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159857.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159857.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159857.hg.1 |
ENST00000291906.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900159857.hg.1 |
NM_013355.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900159857.hg.1 |
TC0900008881.hg.1 |
Clariom D TC |