Assay Details
| Gene Symbol: MRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006039.4 | NP_006030.2 | 16 - 17 | 2877 | 104 | |
| XM_011525543.2 | XP_011523845.1 | 16 - 17 | 2608 | 104 | ||
| GenBank mRNA | AB014609.2 | - | 16 - 17 | 2877 | 104 | |
| AF107292.1 | - | 16 - 17 | 2518 | 104 | ||
| AF134838.1 | - | 16 - 17 | 2599 | 104 | ||
| BC146647.1 | - | 16 - 17 | 2596 | 40147137 | 104 | |
| BC150212.1 | - | 16 - 17 | 2581 | 40148946 | 104 | |
| BC153884.1 | - | 16 - 17 | 2877 | 8860500 | 104 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000758.hg.1
Set Membership: |
|

MRC2
Genomic Map