Assay Details
| Gene Symbol: ABHD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320126.1 | NP_001307055.1 | 2 - 3 | 480 | 102 | |
| XM_005265334.3 | XP_005265391.1 | 2 - 3 | 476 | 102 | ||
| XM_017006930.1 | XP_016862419.1 | 1 - 2 | 187 | 102 | ||
| GenBank mRNA | AK122983.1 | - | 2 - 3 | 172 | 102 | |
| AK225942.1 | - | 3 - 4 | 294 | 102 | ||
| DB171182.1 | - | 2 - 3 | 401 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK122983.1, NM_001320126.1, XM_005265334.3, XM_017006930.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phospholipid catabolic process
negative regulation of cell migration
acylglycerol catabolic process
positive regulation of lipid biosynthetic process
long term synaptic depression
regulation of endocannabinoid signaling pathway
phospholipase activity
acylglycerol lipase activity
phospholipid catabolic process
negative regulation of cell migration
acylglycerol catabolic process
positive regulation of lipid biosynthetic process
long term synaptic depression
regulation of endocannabinoid signaling pathway
phospholipase activity
acylglycerol lipase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000480457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000480457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000480457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000480457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |

ABHD6
Genomic Map