Assay Details
| Gene Symbol: ABHD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320126.1 | NP_001307055.1 | 5 - 6 | 896 | 84 | |
| NM_020676.6 | NP_065727.4 | 4 - 5 | 831 | 84 | ||
| XM_005265334.3 | XP_005265391.1 | 5 - 6 | 892 | 84 | ||
| XM_005265335.3 | XP_005265392.1 | 4 - 5 | 801 | 84 | ||
| XM_017006930.1 | XP_016862419.1 | 4 - 5 | 603 | 84 | ||
| GenBank mRNA | AF225418.1 | - | 4 - 5 | 653 | 84 | |
| AK122983.1 | - | 5 - 6 | 588 | 84 | ||
| AK225942.1 | - | 6 - 7 | 1105 | 84 | ||
| AK313168.1 | - | 4 - 5 | 722 | 84 | ||
| BC001698.2 | - | 4 - 5 | 504 | 2966979 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000376.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phospholipid catabolic process
negative regulation of cell migration
acylglycerol catabolic process
positive regulation of lipid biosynthetic process
long term synaptic depression
regulation of endocannabinoid signaling pathway
phospholipase activity
acylglycerol lipase activity
phospholipid catabolic process
negative regulation of cell migration
acylglycerol catabolic process
positive regulation of lipid biosynthetic process
long term synaptic depression
regulation of endocannabinoid signaling pathway
phospholipase activity
acylglycerol lipase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03008165.hg.1 | TC03000376.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03008165.hg.1 | ENST00000295962.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03008165.hg.1 | ENST00000478253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03008165.hg.1 | ENST00000511761.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03008165.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000480457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000480457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300155001.hg.1 | TC0300007606.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000295962.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000463756.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000470440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000475982.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000478253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000480457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000480457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000485900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | ENST00000613521.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007606.hg.1 | NM_020676.6 | Refseq |

ABHD6
Genomic Map