Assay Details
| Gene Symbol: IQSEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011534311.2 | XP_011532613.1 | 1 - 2 | 478 | 132 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011534311.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000473088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000473088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000473088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000474467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000474467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000273221.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000473088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000473088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000473088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000474467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000474467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000613206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | ENST00000618604.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_001134382.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010315.hg.1 | NM_014869.6 | Refseq |

IQSEC1
Genomic Map