Assay Details
| Gene Symbol: GPR160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005247346.3 | XP_005247403.1 | 2 - 3 | 204 | 68 | |
| XM_011512678.2 | XP_011510980.1 | 2 - 3 | 393 | 68 | ||
| XM_017006161.1 | XP_016861650.1 | 2 - 3 | 204 | 68 | ||
| GenBank mRNA | AK311293.1 | - | 2 - 3 | 204 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005247346.3, XM_011512678.2, XM_017006161.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000461049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000461049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000355897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000461049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000461049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000473675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000476668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000478364.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000482813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000485735.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000491740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | ENST00000492924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009459.hg.1 | NM_014373.2 | Refseq |

GPR160
Genomic Map