Assay Details
| Gene Symbol: MLXIPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134541.1 | 10 - 11 | 1727 | 82 | ||
| GenBank mRNA | AF245474.1 | - | 10 - 11 | 1676 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transducin (beta)-like 2 | TBL2 |
WBSCR13, WS-betaTRP |
-17.7kb | Sense | Chr. 7 - 73568944 - 73578683 on NCBI Build GRCh38 |
| Downstream |
VPS37D, ESCRT-I subunit |
VPS37D | WBSCR24 | 70.9kb | Anti-sense | Chr. 7 - 73665346 - 73672110 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MLXIPL
Genomic Map