| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018584.hg.1 |
TC09001494.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018584.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018584.hg.1 |
ENST00000491462.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09018584.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011241.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176555.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176555.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176555.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176555.hg.1 |
ENST00000374228.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176555.hg.1 |
ENST00000491462.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900176555.hg.1 |
NM_033051.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900176555.hg.1 |
TC0900011241.hg.1 |
Clariom D TC |