Assay Details
| Gene Symbol: EXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286441.1 | NP_001273370.1 | 5 - 6 | 714 | 89 | |
| NM_152596.3 | NP_689809.2 | 3 - 4 | 459 | 89 | ||
| XM_006720415.3 | XP_006720478.1 | 4 - 5 | 631 | 89 | ||
| XM_011521298.2 | XP_011519600.1 | 6 - 7 | 944 | 89 | ||
| XM_011521299.2 | XP_011519601.1 | 6 - 7 | 675 | 89 | ||
| XM_011521300.2 | XP_011519602.1 | 3 - 4 | 203 | 89 | ||
| XM_011521301.2 | XP_011519603.1 | 5 - 6 | 500 | 89 | ||
| XM_017021970.1 | XP_016877459.1 | 3 - 4 | 203 | 89 | ||
| XM_017021971.1 | XP_016877460.1 | 3 - 4 | 201 | 89 | ||
| GenBank mRNA | AK292524.1 | - | 3 - 4 | 365 | 89 | |
| AK302864.1 | - | 5 - 6 | 648 | 89 | ||
| BC030628.2 | - | 3 - 4 | 399 | 4827307 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001247.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011317.hg.1 | ENST00000314992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011317.hg.1 | ENST00000458580.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011317.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011317.hg.1 | TC15001247.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011318.hg.1 | ENST00000314992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011318.hg.1 | ENST00000458580.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011318.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011318.hg.1 | TC15001247.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500166236.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166236.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166236.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166236.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166236.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166236.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166236.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166236.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166236.hg.1 | TC1500009143.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000314992.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000458580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000558881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | ENST00000559743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_001286441.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009143.hg.1 | NM_152596.3 | Refseq |

EXD1
Genomic Map