Assay Details
| Gene Symbol: BRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007371.3 | NP_031397.1 | 1 - 2 | 72 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001697.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_007371.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09021849.hg.1 | TC09001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021849.hg.1 | ENST00000303407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021849.hg.1 | ENST00000357885.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021849.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021850.hg.1 | TC09001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021850.hg.1 | ENST00000303407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021850.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021846.hg.1 | TC09001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021846.hg.1 | ENST00000303407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021846.hg.1 | ENST00000357885.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021846.hg.1 | ENST00000371834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021846.hg.1 | ENST00000540795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021846.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181357.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181357.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181357.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181357.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181357.hg.1 | TC0900011872.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |

BRD3
Genomic Map