Assay Details
| Gene Symbol: BRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007371.3 | NP_031397.1 | 4 - 5 | 689 | 135 | |
| XM_006717291.2 | XP_006717354.1 | 4 - 5 | 821 | 135 | ||
| XM_011519052.2 | XP_011517354.1 | 4 - 5 | 1040 | 135 | ||
| XM_017015165.1 | XP_016870654.1 | 3 - 4 | 614 | 135 | ||
| GenBank mRNA | AY513270.1 | - | 3 - 4 | 519 | 135 | |
| BC032124.2 | - | 4 - 5 | 799 | 4856840 | 135 | |
| D26362.1 | - | 4 - 5 | 640 | 135 | ||
| KR633047.1 | - | 3 - 4 | 501 | 135 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001697.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09021841.hg.1 | TC09001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021841.hg.1 | ENST00000303407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021841.hg.1 | ENST00000357885.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021841.hg.1 | ENST00000371834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021841.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021840.hg.1 | TC09001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021840.hg.1 | ENST00000303407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021840.hg.1 | ENST00000357885.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021840.hg.1 | ENST00000371834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021840.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021839.hg.1 | TC09001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021839.hg.1 | ENST00000303407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021839.hg.1 | ENST00000357885.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021839.hg.1 | ENST00000371834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021839.hg.1 | ENST00000540795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021839.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181348.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181348.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181348.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181348.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181348.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181348.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181348.hg.1 | TC0900011872.hg.1 | Clariom D TC |

BRD3
Genomic Map