Assay Details
| Gene Symbol: BRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007371.3 | NP_031397.1 | 5 - 6 | 905 | 88 | |
| XM_006717291.2 | XP_006717354.1 | 5 - 6 | 1037 | 88 | ||
| XM_011519052.2 | XP_011517354.1 | 5 - 6 | 1256 | 88 | ||
| XM_017015165.1 | XP_016870654.1 | 4 - 5 | 830 | 88 | ||
| GenBank mRNA | AK299519.1 | - | 5 - 6 | 940 | 88 | |
| AY513270.1 | - | 4 - 5 | 735 | 88 | ||
| BC032124.2 | - | 5 - 6 | 1015 | 4856840 | 88 | |
| D26362.1 | - | 5 - 6 | 856 | 88 | ||
| KR633047.1 | - | 4 - 5 | 717 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001697.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK299519.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09021837.hg.1 | TC09001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021837.hg.1 | ENST00000303407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021837.hg.1 | ENST00000357885.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021837.hg.1 | ENST00000371834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021837.hg.1 | ENST00000540795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021837.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021838.hg.1 | TC09001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021838.hg.1 | ENST00000303407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021838.hg.1 | ENST00000357885.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021838.hg.1 | ENST00000371834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021838.hg.1 | ENST00000540795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021838.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181346.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181346.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181346.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181346.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181346.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181346.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181346.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181346.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181346.hg.1 | TC0900011872.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181347.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181347.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181347.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181347.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181347.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181347.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181347.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181347.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181347.hg.1 | TC0900011872.hg.1 | Clariom D TC |

BRD3
Genomic Map