Assay Details
| Gene Symbol: BRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007371.3 | NP_031397.1 | 6 - 7 | 1274 | 85 | |
| XM_006717291.2 | XP_006717354.1 | 6 - 7 | 1406 | 85 | ||
| XM_011519052.2 | XP_011517354.1 | 6 - 7 | 1625 | 85 | ||
| XM_017015165.1 | XP_016870654.1 | 5 - 6 | 1199 | 85 | ||
| XM_017015166.1 | XP_016870655.1 | 3 - 4 | 591 | 85 | ||
| GenBank mRNA | AK299519.1 | - | 6 - 7 | 1309 | 85 | |
| AY513270.1 | - | 5 - 6 | 1104 | 85 | ||
| BC032124.2 | - | 6 - 7 | 1384 | 4856840 | 85 | |
| D26362.1 | - | 6 - 7 | 1225 | 85 | ||
| KR633047.1 | - | 5 - 6 | 1086 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001697.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09021833.hg.1 | TC09001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021833.hg.1 | ENST00000303407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021833.hg.1 | ENST00000357885.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021833.hg.1 | ENST00000371834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021833.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021833.hg.1 | ENST00000540795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021833.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021835.hg.1 | TC09001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021835.hg.1 | ENST00000303407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021835.hg.1 | ENST00000357885.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021835.hg.1 | ENST00000371834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021835.hg.1 | ENST00000540795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021835.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021834.hg.1 | TC09001697.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021834.hg.1 | ENST00000303407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021834.hg.1 | ENST00000357885.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021834.hg.1 | ENST00000371834.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021834.hg.1 | ENST00000540795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021834.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000371842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000433041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | ENST00000494743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011872.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181340.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181340.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181340.hg.1 | TC0900011872.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900181343.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181343.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181343.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181343.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181343.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181343.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181343.hg.1 | TC0900011872.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900181342.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181342.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181342.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181342.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181342.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181342.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181342.hg.1 | ENST00000473349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181342.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181342.hg.1 | TC0900011872.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900181344.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181344.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181344.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181344.hg.1 | ENST00000303407.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181344.hg.1 | ENST00000371834.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181344.hg.1 | NM_007371.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181344.hg.1 | TC0900011872.hg.1 | Clariom D TC |

BRD3
Genomic Map