| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009068.hg.1 |
TC09000724.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009068.hg.1 |
ENST00000259339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009068.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009068.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009067.hg.1 |
TC09000724.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009067.hg.1 |
ENST00000259339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009067.hg.1 |
ENST00000437263.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009067.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009067.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009065.hg.1 |
TC09000724.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009065.hg.1 |
ENST00000259339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009065.hg.1 |
ENST00000437263.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009065.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009065.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009064.hg.1 |
TC09000724.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009064.hg.1 |
ENST00000259339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009064.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009064.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009066.hg.1 |
TC09000724.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009066.hg.1 |
ENST00000259339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009066.hg.1 |
ENST00000437263.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009066.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09009066.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
ENST00000488169.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008933.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160555.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160555.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160555.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160555.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160555.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160555.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160555.hg.1 |
TC0900008933.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900160558.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160558.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160558.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160558.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160558.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160558.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160558.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160558.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160558.hg.1 |
TC0900008933.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900160557.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160557.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160557.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160557.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160557.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160557.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160557.hg.1 |
TC0900008933.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900160559.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160559.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160559.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160559.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160559.hg.1 |
ENST00000259339.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160559.hg.1 |
ENST00000427860.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160559.hg.1 |
ENST00000486372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900160559.hg.1 |
NM_014506.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900160559.hg.1 |
TC0900008933.hg.1 |
Clariom D TC |