| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010863.hg.1 |
ENST00000566170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000359782.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379250.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379250.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379250.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379250.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379250.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379250.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379250.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379250.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379250.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379250.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379250.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000379262.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000397350.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606117.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606427.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606427.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606427.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606427.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
ENST00000606427.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001193267.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_001242827.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_012241.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006992.hg.1 |
NM_031244.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010863.hg.1 |
ENST00000566170.1 |
Ensembl |