Assay Details
| Gene Symbol: ITGA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002207.2 | NP_002198.2 | 16 - 17 | 1886 | 91 | |
| GenBank mRNA | D25303.1 | - | 17 - 18 | 1884 | 91 | |
| L24158.1 | - | 16 - 17 | 1728 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000181.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03003434.hg.1 | TC03000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003434.hg.1 | ENST00000264741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003434.hg.1 | ENST00000422441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003434.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003436.hg.1 | TC03000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003436.hg.1 | ENST00000264741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003436.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003438.hg.1 | TC03000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003438.hg.1 | ENST00000264741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003438.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149208.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149208.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149208.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149208.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149208.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149208.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149208.hg.1 | TC0300007051.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149212.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149212.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149212.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149212.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149212.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149212.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149212.hg.1 | TC0300007051.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300149210.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149210.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149210.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149210.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149210.hg.1 | TC0300007051.hg.1 | Clariom D TC |

ITGA9
Genomic Map