Assay Details
| Gene Symbol: ITGA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002207.2 | NP_002198.2 | 20 - 21 | 2289 | 78 | |
| GenBank mRNA | D25303.1 | - | 21 - 22 | 2287 | 78 | |
| L24158.1 | - | 20 - 21 | 2131 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000181.hg.1
Set Membership: |
|

ITGA9
Genomic Map