Assay Details
| Gene Symbol: ITGA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002207.2 | NP_002198.2 | 22 - 23 | 2488 | 78 | |
| GenBank mRNA | D25303.1 | - | 23 - 24 | 2486 | 78 | |
| L24158.1 | - | 22 - 23 | 2330 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000181.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03003444.hg.1 | TC03000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003444.hg.1 | ENST00000264741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003444.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149218.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149218.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149218.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149218.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149218.hg.1 | TC0300007051.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |

ITGA9
Genomic Map