Assay Details
| Gene Symbol: ITGA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002207.2 | NP_002198.2 | 8 - 9 | 948 | 80 | |
| GenBank mRNA | BC030198.1 | - | 8 - 9 | 1155 | 5199296 | 80 |
| D25303.1 | - | 9 - 10 | 946 | 80 | ||
| L24158.1 | - | 8 - 9 | 790 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000181.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003427.hg.1 | TC03000181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003427.hg.1 | ENST00000264741.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003427.hg.1 | ENST00000422441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03003427.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149201.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149201.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149201.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149201.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149201.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300149201.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300149201.hg.1 | TC0300007051.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000264741.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000411817.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000422441.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | ENST00000461533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007051.hg.1 | NM_002207.2 | Refseq |

ITGA9
Genomic Map