Assay Details
Gene Symbol: KREMEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032045.4 | NP_114434.3 | 9 - 10 | 1467 | 80 | |
GenBank mRNA | AK056425.1 | - | 9 - 10 | 1427 | 80 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000191.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22003511.hg.1 | ENST00000327813.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22003511.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22003511.hg.1 | TC22000191.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000453585.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000453585.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000453585.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000453585.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000474001.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000474001.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000474001.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000474001.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000479755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | ENST00000479755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22003505.hg.1 | ENST00000327813.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22003505.hg.1 | ENST00000400335.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22003505.hg.1 | ENST00000400338.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22003505.hg.1 | ENST00000407188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22003505.hg.1 | ENST00000479755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22003505.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22003505.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22003505.hg.1 | TC22000191.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2200168411.hg.1 | TC2200009248.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200168411.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168411.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200168411.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168411.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | TC2200009248.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | ENST00000479755.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | ENST00000479755.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200168405.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200168410.hg.1 | TC2200009248.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200168410.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168410.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200168410.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200168410.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000327813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000400335.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000407188.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000453585.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000453585.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000453585.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000453585.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000474001.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000474001.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000474001.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000474001.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000479755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | ENST00000479755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_001039570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009248.hg.1 | NM_032045.4 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 184815 | - | - |
