Assay Details
| Gene Symbol: MGAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011530190.2 | XP_011528492.1 | 1 | 2719 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100506472 | LOC100506472 | -34.7kb | Anti-sense | Chr. 22 - 39432160 - 39437128 on NCBI Build GRCh38 | |
| Downstream | MGAT3 antisense RNA 1 | MGAT3-AS1 | TapSAKI | 3.9kb | Sense | Chr. 22 - 39475807 - 39476822 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011530190.2 transcripts.
Set Membership: |
|

MGAT3
Genomic Map