Assay Details
| Gene Symbol: TP63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB042841.1 | - | 1 - 2 | 43 | 75 | |
| KP881231.1 | - | 1 - 2 | 48 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TP63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB042841.1 | - | 1 - 2 | 43 | 75 | |
| KP881231.1 | - | 1 - 2 | 48 | 75 | ||
Set Membership: |
|