Assay Details
| Gene Symbol: MVB12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033446.2 | NP_258257.1 | 6 - 7 | 753 | 81 | |
| XM_005252297.1 | XP_005252354.1 | 6 - 7 | 788 | 81 | ||
| XM_017015275.1 | XP_016870764.1 | 6 - 7 | 753 | 81 | ||
| XM_017015276.1 | XP_016870765.1 | 6 - 7 | 788 | 81 | ||
| XM_017015277.1 | XP_016870766.1 | 6 - 7 | 753 | 81 | ||
| XM_017015279.1 | XP_016870768.1 | 5 - 6 | 608 | 81 | ||
| GenBank mRNA | AK024432.1 | - | 6 - 7 | 748 | 81 | |
| AK024441.1 | - | 3 - 4 | 275 | 81 | ||
| AK293740.1 | - | 6 - 7 | 747 | 81 | ||
| AK307473.1 | - | 5 - 6 | 585 | 81 | ||
| BC000122.1 | - | 6 - 7 | 728 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000654.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007590.hg.1 | TC09000654.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007590.hg.1 | ENST00000361171.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007590.hg.1 | ENST00000436593.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007590.hg.1 | ENST00000535766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007590.hg.1 | ENST00000545391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007590.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007590.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007593.hg.1 | TC09000654.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007593.hg.1 | ENST00000361171.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007593.hg.1 | ENST00000436593.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007593.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158550.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158550.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158550.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158550.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158550.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158550.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158550.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158550.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158550.hg.1 | TC0900008776.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158547.hg.1 | TC0900008776.hg.1 | Clariom D TC |

MVB12B
Genomic Map