Assay Details
| Gene Symbol: MVB12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033446.2 | NP_258257.1 | 7 - 8 | 846 | 95 | |
| XM_005252297.1 | XP_005252354.1 | 7 - 8 | 881 | 95 | ||
| GenBank mRNA | AK024441.1 | - | 4 - 5 | 368 | 95 | |
| AK293740.1 | - | 7 - 8 | 840 | 95 | ||
| AK307473.1 | - | 6 - 7 | 678 | 95 | ||
| BC000122.1 | - | 7 - 8 | 821 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000654.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09007597.hg.1 | TC09000654.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007597.hg.1 | ENST00000361171.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007597.hg.1 | ENST00000436593.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007597.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007597.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000402437.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000470567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489570.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | ENST00000489637.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_001011703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008776.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158554.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158554.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158554.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158554.hg.1 | ENST00000361171.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158554.hg.1 | ENST00000485886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158554.hg.1 | NM_033446.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158554.hg.1 | TC0900008776.hg.1 | Clariom D TC |

MVB12B
Genomic Map