Assay Details
| Gene Symbol: SETX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015046.5 | NP_055861.3 | 11 - 12 | 5552 | 80 | |
| XM_005272171.1 | XP_005272228.1 | 11 - 12 | 5553 | 80 | ||
| XM_005272172.2 | XP_005272229.1 | 12 - 13 | 5651 | 80 | ||
| XM_005272173.2 | XP_005272230.1 | 11 - 12 | 5689 | 80 | ||
| XM_011518404.2 | XP_011516706.1 | 12 - 13 | 5693 | 80 | ||
| XM_011518405.2 | XP_011516707.1 | 11 - 12 | 5711 | 80 | ||
| XM_011518406.2 | XP_011516708.1 | 11 - 12 | 5553 | 80 | ||
| XM_011518407.1 | XP_011516709.1 | 11 - 12 | 5553 | 80 | ||
| XM_011518408.2 | XP_011516710.1 | 11 - 12 | 5553 | 80 | ||
| XM_017014495.1 | XP_016869984.1 | 11 - 12 | 5624 | 80 | ||
| XM_017014496.1 | XP_016869985.1 | 2 - 3 | 220 | 80 | ||
| XM_017014497.1 | XP_016869986.1 | 2 - 3 | 589 | 80 | ||
| GenBank mRNA | AB014525.2 | - | 9 - 10 | 5329 | 80 | |
| AK001456.1 | - | 1 - 2 | 88 | 80 | ||
| AK226007.1 | - | 2 - 3 | 138 | 80 | ||
| AY362728.1 | - | 9 - 10 | 5376 | 80 | ||
| BC032600.2 | - | 2 - 3 | 129 | 80 | ||
| BC032622.1 | - | 2 - 3 | 430 | 80 | ||
| BC078166.1 | - | 1 - 2 | 89 | 80 | ||
| BC137350.1 | - | 10 - 11 | 5486 | 9021348 | 80 | |
| BX537849.1 | - | 11 - 12 | 5537 | 80 | ||
| BX538166.1 | - | 11 - 12 | 5555 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001673.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK001456.1,XM_017014496.1,XM_017014497.1 transcripts.
Set Membership: |
|

SETX
Genomic Map