Assay Details
| Gene Symbol: FNDC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135095.1 | NP_001128567.1 | 15 - 16 | 1885 | 124 | |
| NM_022763.3 | NP_073600.3 | 15 - 16 | 1958 | 124 | ||
| XM_017007062.1 | XP_016862551.1 | 14 - 15 | 1656 | 124 | ||
| XM_017007063.1 | XP_016862552.1 | 10 - 11 | 1142 | 124 | ||
| XM_017007064.1 | XP_016862553.1 | 15 - 16 | 1938 | 124 | ||
| GenBank mRNA | AB098597.1 | - | 14 - 15 | 1843 | 124 | |
| AK075220.1 | - | 5 - 6 | 580 | 124 | ||
| AK127826.1 | - | 15 - 16 | 1938 | 124 | ||
| AK223599.1 | - | 15 - 16 | 1989 | 124 | ||
| AK314478.1 | - | 15 - 16 | 1885 | 124 | ||
| AY358146.1 | - | 8 - 9 | 831 | 124 | ||
| BC033635.2 | - | 3 - 4 | 433 | 124 | ||
| BC039297.1 | - | 15 - 16 | 1849 | 5297602 | 124 | |
| BX648340.1 | - | 15 - 16 | 1880 | 124 | ||
More Information
Set Membership: |
|

FNDC3B
Genomic Map