Assay Details
| Gene Symbol: NMNAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015039.3 | NP_055854.1 | 4 - 5 | 555 | 75 | |
| NM_170706.3 | NP_733820.1 | 4 - 5 | 443 | 75 | ||
| GenBank mRNA | AB007948.1 | - | 4 - 5 | 423 | 75 | |
| AF288395.1 | - | 4 - 5 | 660 | 75 | ||
| AK291390.1 | - | 6 - 7 | 555 | 75 | ||
| BC020998.2 | - | 4 - 5 | 443 | 2824280 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003614.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01055592.hg.1 | TC01003614.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055592.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055592.hg.1 | ENST00000294868.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055592.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055592.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055592.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000464047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016649.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226300.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226300.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226300.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226300.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226300.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226300.hg.1 | ENST00000287713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226300.hg.1 | ENST00000294868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226300.hg.1 | ENST00000473046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226300.hg.1 | NM_015039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226300.hg.1 | NM_170706.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226300.hg.1 | TC0100016649.hg.1 | Clariom D TC |

NMNAT2
Genomic Map