| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038812.hg.1 |
TC02002467.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038812.hg.1 |
ENST00000263636.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038812.hg.1 |
ENST00000504764.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038812.hg.1 |
ENST00000554112.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038812.hg.1 |
NM_001198759.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038812.hg.1 |
NM_002349.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200207131.hg.1 |
ENST00000263636.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207131.hg.1 |
ENST00000504764.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207131.hg.1 |
NM_001198759.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200207131.hg.1 |
NM_002349.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200207131.hg.1 |
ENST00000263636.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207131.hg.1 |
ENST00000504764.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207131.hg.1 |
NM_001198759.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200207131.hg.1 |
NM_002349.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200207131.hg.1 |
TC0200014728.hg.1 |
Clariom D TC |