Assay Details
| Gene Symbol: DDTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK298326.1 | - | 2 - 3 | 303 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000350608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000398344.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000403754.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000404092.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000428792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000430101.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000444947.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | ENST00000506097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008203.hg.1 | NM_001084392.1 | Refseq |

DDTL
Genomic Map