Assay Details
| Gene Symbol: DUSP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007240.2 | NP_009171.1 | 3 - 4 | 607 | 138 | |
| XM_005244862.2 | XP_005244919.1 | 3 - 4 | 696 | 138 | ||
| GenBank mRNA | AF119226.1 | - | 3 - 4 | 584 | 138 | |
| BC006286.1 | - | 3 - 4 | 605 | 3958403 | 138 | |
| BT006633.1 | - | 3 - 4 | 573 | 138 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001413.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
dephosphorylation
positive regulation of glucokinase activity
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
zinc ion binding
phosphatase activity
kinase binding
cellular protein modification process
dephosphorylation
positive regulation of glucokinase activity
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
zinc ion binding
phosphatase activity
kinase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022114.hg.1 | TC01001413.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022114.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022114.hg.1 | ENST00000484291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022114.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011839.hg.1 | ENST00000612874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011839.hg.1 | ENST00000612874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011839.hg.1 | ENST00000612874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011839.hg.1 | ENST00000612874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022113.hg.1 | TC01001413.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022113.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022113.hg.1 | ENST00000484291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022113.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176368.hg.1 | TC0100010368.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100176369.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176369.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176369.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176369.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176369.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176369.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176369.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176369.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176369.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176369.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176369.hg.1 | TC0100010368.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011839.hg.1 | ENST00000612874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011839.hg.1 | ENST00000612874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011839.hg.1 | ENST00000612874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011839.hg.1 | ENST00000612874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |

DUSP12
Genomic Map