Assay Details
| Gene Symbol: REN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000537.3 | NP_000528.1 | 3 - 4 | 416 | 62 | |
| GenBank mRNA | BC033474.1 | - | 3 - 4 | 398 | 5188566 | 62 |
| BC047752.1 | - | 3 - 4 | 401 | 5590211 | 62 | |
| BP321283.1 | - | 3 - 4 | 416 | 62 | ||
| CR536498.1 | - | 3 - 4 | 372 | 62 | ||
| X00063.1 | - | 1 - 2 | 51 | 62 | ||
More Information
Set Membership: |
|

REN
Genomic Map