Assay Details
| Gene Symbol: POLH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291970.1 | NP_001278899.1 | 2 - 3 | 442 | 85 | |
| NM_006502.2 | NP_006493.1 | 2 - 3 | 442 | 85 | ||
| XM_011514698.2 | XP_011513000.1 | 2 - 3 | 434 | 85 | ||
| GenBank mRNA | AB024313.1 | - | 2 - 3 | 375 | 85 | |
| AF158185.1 | - | 2 - 3 | 303 | 85 | ||
| AK023893.1 | - | 2 - 3 | 433 | 85 | ||
| AK294430.1 | - | 2 - 3 | 423 | 85 | ||
| BC015742.1 | - | 2 - 3 | 286 | 4872554 | 85 | |
| DC416114.1 | - | 2 - 3 | 443 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000605.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK294430.1,XM_011514698.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA synthesis involved in DNA repair
DNA replication
DNA repair
regulation of DNA repair
pyrimidine dimer repair
response to UV-C
translesion synthesis
error-free translesion synthesis
cellular response to UV-C
damaged DNA binding
DNA-directed DNA polymerase activity
protein binding
metal ion binding
DNA synthesis involved in DNA repair
DNA replication
DNA repair
regulation of DNA repair
pyrimidine dimer repair
response to UV-C
translesion synthesis
error-free translesion synthesis
cellular response to UV-C
damaged DNA binding
DNA-directed DNA polymerase activity
protein binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06007234.hg.1 | TC06000605.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007234.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007234.hg.1 | ENST00000372236.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007234.hg.1 | ENST00000535400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06007234.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000443535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000443535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | ENST00000443535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000443535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000443535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | ENST00000443535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008099.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | ENST00000443535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | ENST00000372226.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | ENST00000372236.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | ENST00000443535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | NM_001291969.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | NM_001291970.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | NM_006502.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600156589.hg.1 | TC0600008099.hg.1 | Clariom D TC |

POLH
Genomic Map