Assay Details
| Gene Symbol: DNAJA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005880.3 | NP_005871.1 | 6 - 7 | 916 | 74 | |
| GenBank mRNA | AJ001309.1 | - | 6 - 7 | 826 | 74 | |
| AK313031.1 | - | 6 - 7 | 864 | 74 | ||
| BC013044.2 | - | 6 - 7 | 868 | 3505399 | 74 | |
| BC015809.2 | - | 6 - 7 | 869 | 3922477 | 74 | |
| Y13350.1 | - | 4 - 5 | 511 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001087.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000569553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000569553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000569553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000569553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015267.hg.1 | ENST00000317089.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015267.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015267.hg.1 | TC16001087.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600178450.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178450.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178450.hg.1 | ENST00000569553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178450.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178450.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178450.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178450.hg.1 | ENST00000569553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178450.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178450.hg.1 | TC1600010179.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000317089.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000563158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000569553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000569553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000569553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000569553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | ENST00000617000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010179.hg.1 | NM_005880.3 | Refseq |

DNAJA2
Genomic Map