Assay Details
| Gene Symbol: SLC1A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003038.4 | NP_003029.2 | 4 - 5 | 1085 | 87 | |
| XM_006712079.2 | XP_006712142.1 | 4 - 5 | 451 | 87 | ||
| GenBank mRNA | AB026689.1 | - | 4 - 5 | 823 | 87 | |
| AK126636.1 | - | 3 - 4 | 2275 | 87 | ||
| AK223486.1 | - | 4 - 5 | 1047 | 87 | ||
| AK298472.1 | - | 5 - 6 | 996 | 87 | ||
| AK313052.1 | - | 4 - 5 | 1049 | 87 | ||
| AK315986.1 | - | 5 - 6 | 1007 | 87 | ||
| BC026216.1 | - | 4 - 5 | 986 | 4420405 | 87 | |
| BC072423.1 | - | 4 - 5 | 1045 | 6186177 | 87 | |
| DA218921.1 | - | 4 - 5 | 453 | 87 | ||
| L14595.1 | - | 4 - 5 | 985 | 87 | ||
| L19444.1 | - | 4 - 5 | 964 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000374.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
amino acid transport
glutamine transport
L-alanine transport
L-cystine transport
proline transport
L-serine transport
threonine transport
hydroxyproline transport
synaptic transmission, glutamatergic
proline transmembrane transport
cognition
mitophagy in response to mitochondrial depolarization
chloride transmembrane transport
chloride channel activity
amino acid transmembrane transporter activity
L-alanine transmembrane transporter activity
L-cystine transmembrane transporter activity
L-glutamine transmembrane transporter activity
L-proline transmembrane transporter activity
L-serine transmembrane transporter activity
L-threonine transmembrane transporter activity
symporter activity
L-hydroxyproline transmembrane transporter activity
amino acid transport
glutamine transport
L-alanine transport
L-cystine transport
proline transport
L-serine transport
threonine transport
hydroxyproline transport
synaptic transmission, glutamatergic
proline transmembrane transport
cognition
mitophagy in response to mitochondrial depolarization
chloride transmembrane transport
chloride channel activity
amino acid transmembrane transporter activity
L-alanine transmembrane transporter activity
L-cystine transmembrane transporter activity
L-glutamine transmembrane transporter activity
L-proline transmembrane transporter activity
L-serine transmembrane transporter activity
L-threonine transmembrane transporter activity
symporter activity
L-hydroxyproline transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02005678.hg.1 | TC02000374.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005678.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005678.hg.1 | ENST00000448784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005678.hg.1 | ENST00000493121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005678.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005679.hg.1 | TC02000374.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005679.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005679.hg.1 | ENST00000448784.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005679.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153979.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153979.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153979.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153979.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153979.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153979.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153979.hg.1 | TC0200007821.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200153978.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153978.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153978.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153978.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153978.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153978.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153978.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153978.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153978.hg.1 | TC0200007821.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000234256.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000471551.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000480594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000493121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | ENST00000531327.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_001193493.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007821.hg.1 | NM_003038.4 | Refseq |

SLC1A4
Genomic Map