Assay Details
| Gene Symbol: MEP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308171.1 | NP_001295100.1 | 11 - 12 | 1630 | 89 | |
| NM_005925.2 | NP_005916.2 | 11 - 12 | 1630 | 89 | ||
| XM_011526013.2 | XP_011524315.1 | 10 - 11 | 1396 | 89 | ||
| XM_011526014.2 | XP_011524316.1 | 9 - 10 | 1243 | 89 | ||
| GenBank mRNA | AY695931.1 | - | 11 - 12 | 1641 | 89 | |
| BC136559.1 | - | 11 - 12 | 1608 | 9020549 | 89 | |
| BC144244.1 | - | 11 - 12 | 1608 | 9052764 | 89 | |
| X81333.1 | - | 11 - 12 | 1629 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000135.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001513.hg.1 | ENST00000269202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001513.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18001513.hg.1 | TC18000135.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1800148184.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800148184.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800148184.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800148184.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800148184.hg.1 | TC1800007037.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800148184.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800148184.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800148184.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800148184.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000269202.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000579919.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581184.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | ENST00000581447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_001308171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007037.hg.1 | NM_005925.2 | Refseq |

MEP1B
Genomic Map