Assay Details
| Gene Symbol: RHBDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017821.4 | NP_060291.2 | 1 - 2 | 146 | 55 | |
| GenBank mRNA | BX951305.1 | - | 1 - 2 | 101 | 55 | |
| DB036310.1 | - | 1 - 2 | 146 | 55 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002534.hg
Additional Information:
This assay is in the 5'UTR region of the NM_017821.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01039483.hg.1 | TC01002534.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039483.hg.1 | ENST00000372990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039483.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039483.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039479.hg.1 | TC01002534.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039479.hg.1 | ENST00000289248.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039479.hg.1 | ENST00000372985.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039479.hg.1 | ENST00000372990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039479.hg.1 | ENST00000538156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039479.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039479.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202991.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202991.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202991.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202991.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202991.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202991.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202991.hg.1 | TC0100013778.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100202986.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202986.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202986.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202986.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202986.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202986.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202986.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202986.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202986.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202986.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202986.hg.1 | TC0100013778.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100202993.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202993.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202993.hg.1 | TC0100013778.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100202987.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202987.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202987.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202987.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202987.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202987.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202987.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202987.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202987.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202987.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202987.hg.1 | TC0100013778.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000289248.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000372990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | ENST00000540558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_001304746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013778.hg.1 | NM_017821.4 | Refseq |

RHBDL2
Genomic Map