Assay Details
| Gene Symbol: CFAP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144872.1 | NP_001138344.1 | 4 - 5 | 466 | 94 | |
| XM_011538327.2 | XP_011536629.1 | 4 - 5 | 1162 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000888.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12011740.hg.1 | ENST00000335621.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011740.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011740.hg.1 | TC12000888.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000550918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000550918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000550918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166380.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166380.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166380.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166380.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166380.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166380.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166380.hg.1 | TC1200008929.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000335621.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000550918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000550918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000550918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | ENST00000551256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | NM_001144872.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008929.hg.1 | NM_001144872.2 | Refseq |

CFAP73
Genomic Map