Assay Details
| Gene Symbol: CORO2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003389.3 | NP_003380.3 | 3 - 4 | 585 | 72 | |
| NM_052820.3 | NP_438171.1 | 3 - 4 | 435 | 72 | ||
| XM_011518986.2 | XP_011517288.1 | 3 - 4 | 762 | 72 | ||
| GenBank mRNA | AK292788.1 | - | 3 - 4 | 379 | 72 | |
| BC000010.1 | - | 2 - 3 | 323 | 3504229 | 72 | |
| BC011690.2 | - | 3 - 4 | 401 | 3350035 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001404.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09017484.hg.1 | TC09001404.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017484.hg.1 | ENST00000343933.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017484.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017484.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017484.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174744.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174744.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174744.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174744.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174744.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174744.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174744.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174744.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174744.hg.1 | TC0900010969.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |

CORO2A
Genomic Map