Assay Details
Gene Symbol: CORO2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003389.3 | NP_003380.3 | 7 - 8 | 1139 | 66 | |
NM_052820.3 | NP_438171.1 | 7 - 8 | 989 | 66 | ||
XM_011518986.2 | XP_011517288.1 | 7 - 8 | 1316 | 66 | ||
GenBank mRNA | AK292788.1 | - | 7 - 8 | 933 | 66 | |
BC000010.1 | - | 6 - 7 | 877 | 3504229 | 66 | |
BC011690.2 | - | 7 - 8 | 955 | 3350035 | 66 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001404.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09017480.hg.1 | TC09001404.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09017480.hg.1 | ENST00000343933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09017480.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09017480.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09017480.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174740.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174740.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174740.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174740.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174740.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174740.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900174740.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174740.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900174740.hg.1 | TC0900010969.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000343933.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | ENST00000375077.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_003389.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010969.hg.1 | NM_052820.3 | Refseq |
